Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRIQ4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191385.1 | NP_001178314.1 | 3 - 4 | 1326 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Bos taurus Probe spans exons |