Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191403.1 | NP_001178332.1 | 2 - 3 | 286 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |