Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191427.1 | NP_001178356.1 | 3 - 4 | 342 | 118 | |
GenBank mRNA | BC102544.1 | - | 3 - 4 | 231 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |