Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244204.1 | NP_001231133.1 | 7 - 8 | 967 | 102 | |
GenBank mRNA | EV744049.1 | - | 1 - 2 | 148 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Bos taurus Probe spans exons |