Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244214.1 | NP_001231143.1 | 3 - 4 | 216 | 68 | |
GenBank mRNA | BC114780.1 | - | 3 - 4 | 175 | 68 | |
EW682951.1 | - | 3 - 4 | 216 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |