Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECT2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191539.1 | NP_001178468.1 | 11 - 12 | 1581 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |