Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191541.1 | NP_001178470.1 | 19 - 20 | 3006 | 90 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Bos taurus Probe spans exons |