Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRHPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192037.3 | NP_001178966.1 | 7 - 8 | 760 | 106 | |
GenBank mRNA | BC120229.1 | - | 7 - 8 | 765 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |