Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCN2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192074.2 | NP_001179003.1 | 3 - 4 | 544 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Bos taurus Probe spans exons |