Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192080.2 | NP_001179009.2 | 3 - 4 | 411 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |