Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192088.3 | NP_001179017.1 | 8 - 9 | 820 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Bos taurus Probe spans exons |