Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB5D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192138.2 | NP_001179067.2 | 3 - 4 | 510 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |