Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192139.1 | NP_001179068.1 | 6 - 7 | 915 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Bos taurus Probe spans exons |