Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192153.3 | NP_001179082.2 | 3 - 4 | 602 | 73 | |
GenBank mRNA | BC149556.1 | - | 3 - 4 | 486 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |