Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSC4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192192.2 | NP_001179121.2 | 3 - 4 | 503 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |