Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192218.3 | NP_001179147.1 | 7 - 8 | 1147 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |