Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192266.1 | NP_001179195.1 | 1 - 2 | 155 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |