Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PES1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001354818.1 | NP_001341747.1 | 2 - 3 | 192 | 103 | |
GenBank mRNA | BC118072.1 | - | 2 - 3 | 151 | 103 | |
DT853420.1 | - | 2 - 3 | 129 | 103 | ||
EH377693.1 | - | 2 - 3 | 128 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |