Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192321.1 | NP_001179250.1 | 9 - 10 | 1487 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |