Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192324.1 | NP_001179253.1 | 3 - 4 | 1175 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |