Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC45A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192339.1 | NP_001179268.1 | 1 - 2 | 92 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |