Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192342.1 | NP_001179271.1 | 1 - 2 | 59 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Probe spans exons |