Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192373.1 | NP_001179302.1 | 1 - 2 | 412 | 67 | |
GenBank mRNA | BC142311.1 | - | 1 - 2 | 413 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |