Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192425.1 | NP_001179354.1 | 1 - 2 | 75 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |