Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192444.2 | NP_001179373.2 | 5 - 6 | 639 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |