Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHYKPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192899.2 | NP_001179828.1 | 10 - 11 | 1166 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |