Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHYKPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192899.2 | NP_001179828.1 | 1 - 2 | 62 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |