Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192933.1 | NP_001179862.1 | 6 - 7 | 1041 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |