Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTDSPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193081.2 | NP_001180010.2 | 3 - 4 | 282 | 69 | |
GenBank mRNA | CK776230.1 | - | 3 - 4 | 282 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |