Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193132.1 | NP_001180061.1 | 3 - 4 | 399 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |