Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PJVK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193183.1 | NP_001180112.1 | 2 - 3 | 404 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |