Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANP32A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195019.1 | NP_001181948.1 | 4 - 5 | 674 | 167 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |