Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195024.2 | NP_001181953.1 | 1 - 2 | 171 | 123 | |
GenBank mRNA | BC102104.1 | - | 1 - 2 | 173 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |