Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASTKD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024528.2 | NP_001019699.2 | 3 - 4 | 1739 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |