Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRFAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035344.2 | NP_001030421.1 | 1 - 2 | 550 | 64 | |
GenBank mRNA | BC102898.1 | - | 1 - 2 | 551 | 64 | |
BC151429.1 | - | 1 - 2 | 574 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |