Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075201.1 | NP_001068669.1 | 6 - 7 | 605 | 71 | |
GenBank mRNA | BC118392.1 | - | 6 - 7 | 605 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |