Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVA1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199027.1 | NP_001185956.1 | 3 - 4 | 475 | 89 | |
GenBank mRNA | BC126735.1 | - | 2 - 3 | 382 | 89 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Bos taurus Probe spans exons |