Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM71E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205375.2 | NP_001192304.1 | 1 - 2 | 93 | 88 | |
GenBank mRNA | BC109767.1 | - | 1 - 2 | 94 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |