Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205376.2 | NP_001192305.1 | 2 - 3 | 110 | 84 | |
GenBank mRNA | BC103023.1 | - | 2 - 3 | 101 | 84 | |
EH182364.1 | - | 2 - 3 | 110 | 84 | ||
EV683183.1 | - | 2 - 3 | 110 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |