Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TASOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205393.2 | NP_001192322.1 | 6 - 7 | 881 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |