Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205400.2 | NP_001192329.1 | 5 - 6 | 646 | 65 | |
GenBank mRNA | AF491875.1 | - | 2 - 3 | 263 | 65 | |
BC148117.1 | - | 5 - 6 | 647 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |