Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195027.1 | NP_001181956.1 | 1 - 2 | 149 | 101 | |
GenBank mRNA | BC102192.1 | - | 1 - 2 | 103 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |