Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM241A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076961.2 | NP_001070429.1 | 2 | 818 | 92 | |
GenBank mRNA | BC118416.1 | - | 2 | 820 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |