Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_132766.1 | 4 - 5 | 685 | 100 | ||
GenBank mRNA | BC120438.1 | - | 4 - 5 | 671 | 100 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | acyltransferase like 1B-like pseudogene | LOC530756 | -16.3kb | Sense | Chr. 3 - 51331071 - 51333472 on NCBI Build ARS-UCD1.2 | |
Downstream | bromodomain testis associated | BRDT | 30.4kb | Sense | Chr. 3 - 51387227 - 51406793 on NCBI Build ARS-UCD1.2 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |