Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270317.4 | NP_001257246.2 | 22 - 23 | 6540 | 61 | |
NM_001373583.1 | NP_001360775.1 | 22 - 23 | 6507 | 61 | ||
NM_001381151.1 | NP_001368758.1 | 22 - 23 | 6523 | 61 | ||
NM_001381152.1 | NP_001368759.1 | 22 - 23 | 6523 | 61 | ||
NM_001381153.1 | NP_001368756.1 | 22 - 23 | 6490 | 61 | ||
NM_001381154.1 | NP_001368757.1 | 22 - 23 | 6490 | 61 | ||
NM_001381155.1 | NP_001368146.1 | 22 - 23 | 6517 | 61 | ||
NM_001381156.2 | NP_001368760.1 | 22 - 23 | 6513 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |