Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sprr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078054.7 | NP_510455.2 | 4 - 5 | 385 | 95 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |