Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078071.7 | NP_510472.2 | 3 - 4 | 768 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |