Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nac-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078072.5 | NP_510473.2 | 8 - 9 | 1273 | 66 | |
GenBank mRNA | AY090484.1 | - | 8 - 9 | 1283 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |