Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mlt-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078110.4 | NP_510511.1 | 9 - 10 | 1662 | 91 | |
NM_078111.4 | NP_510512.1 | 9 - 10 | 1604 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |