Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270326.3 | NP_001257255.1 | 3 - 4 | 324 | 64 | |
NM_001350352.4 | NP_001337313.1 | 1 - 2 | 319 | 64 | ||
NM_001350353.4 | NP_001337312.1 | 1 - 2 | 319 | 64 | ||
NM_001350354.3 | NP_001337317.1 | 2 - 3 | 148 | 64 | ||
NM_001350355.3 | NP_001337316.1 | 2 - 3 | 70 | 64 | ||
NM_001350356.3 | NP_001337319.1 | 5 | 500 | 64 | ||
NM_001350357.3 | NP_001337318.1 | 2 - 3 | 109 | 64 | ||
NM_001350358.2 | NP_001337320.1 | 3 - 4 | 324 | 64 | ||
NM_001375177.1 | NP_001361907.1 | 3 - 4 | 326 | 64 | ||
GenBank mRNA | Z19122.1 | - | 345 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |